2-Chloroethanesulfonic acid sodium salt hydrate ∼13% in water

Supplier: TCI America

C1110-25G C1110-500G
TCC1110-025GEA 88.48 CAD
TCC1110-025G TCC1110-500G
2-Chloroethanesulfonic acid sodium salt hydrate ∼13% in water
2-Chloroethanesulfonic acid sodium salt

CAS Number: 15484-44-3
MDL Number: MFCD00007531
Molecular Formula: C2H5ClO3S
Molecular Weight: 166.55
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: 2-Chloroethanesulfonic Acid Sodium Salt

Formula: C₂H₄ClNaO₃S
MDL Number: MFCD00007531
CAS Number: 15484-44-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfcf973792cc0c4b48f18816e03f134d84714b4e&vid=ab8d62ddca36ed021e2297abafc66330ca99c74e&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR