4-Chloro-1-methylpiperidine hydrochloride ≥98.0%

Supplier: TCI America

C1085-25G C1085-100G
TCC1085-025GEA 37.92 CAD
TCC1085-025G TCC1085-100G
4-Chloro-1-methylpiperidine hydrochloride ≥98.0%
4-Chloro-1-methylpiperidine hydrochloride

CAS Number: 5382-23-0
MDL Number: MFCD00012834
Molecular Formula: C6H12ClN
Molecular Weight: 170.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 162

Formula: C₆H₁₃Cl₂N
MW: 170.08 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00012834
CAS Number: 5382-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eee9d54e1d010e43aaa64f614ebc65f5d89cd07f&vid=eee9d54e1d010e43aaa64f614ebc65f5d89cd07f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR