N-Cyclohexylethanolamine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C1082-500G
C1082-25G
TCC1082-500GEA
573.54
CAD
TCC1082-500G
TCC1082-025G
N-Cyclohexylethanolamine ≥98.0% (by titrimetric analysis)
N-Cyclohexylethanolamine
CAS Number: 2842-38-8
MDL Number: MFCD00021326
Molecular Formula: C8H17NO
Molecular Weight: 143.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Flash Point (°C): 121
Freezing point (°C): 36
Other Product Names / Synonyms: N-(2-Hydroxyethyl)cyclohexylamine, 2-(Cyclohexylamino)ethanol
Formula:
C₈H₁₇NO Flash Pt: 121 °C |
MDL Number:
MFCD00021326 CAS Number: 2842-38-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...