3-Chloropropylamine hydrochloride ≥97.0%

Supplier: TCI America

Synonyms: 3-Chloropropan-1-amine hydrochloride

C1062-25G C1062-100G
TCC1062-025GEA 37.92 CAD
TCC1062-025G TCC1062-100G
3-Chloropropylamine hydrochloride ≥97.0%
3-Chloropropylamine hydrochloride

CAS Number: 6276-54-6
MDL Number: MFCD00012913
Molecular Formula: C3H8ClN
Molecular Weight: 130.01
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 149

Formula: C₃H₈ClN
MW: 130.01 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00012913
CAS Number: 6276-54-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=009cf55dc674e088072d45bc3015b6e2cabbd23e&vid=009cf55dc674e088072d45bc3015b6e2cabbd23e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR