Cyclopropyl phenyl sulfide ≥98.0%
Supplier: TCI America
Synonyms:
Cyclopropyl(phenyl)sulphane, Cyclopropyl(phenyl)sulfane
C1014-25G
C1014-5G
TCC1014-25GEA
376.99
CAD
TCC1014-25G
TCC1014-5G
Cyclopropyl phenyl sulfide ≥98.0%
Cyclopropyl phenyl sulphide
CAS Number: 14633-54-6
MDL Number: MFCD00009684
Molecular Formula: C9H10S
Molecular Weight: 150.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 91
Specific Gravity (20/20): 1.06
Formula:
C₉H₁₀S Boiling Pt: 90 °C Density: 1.06 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00009684 CAS Number: 14633-54-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...