3-Chloro-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C0988-25G
C0988-5G
TCC0988-25GEA
221.52
CAD
TCC0988-25G
TCC0988-5G
3-Chloro-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
3-Chloro-2-nitrobenzoic acid
CAS Number: 4771-47-5
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 239
Formula:
C₇H₄ClNO₄ Melting Pt: 239 °C |
MDL Number:
MFCD00007087 CAS Number: 4771-47-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...