N-Chloroacetylglycine ≥99.0% (by titrimetric analysis)
Supplier: TCI America
C0980-1G
C0980-25G
TCC0980-001GEA
71.1
CAD
TCC0980-001G
TCC0980-025G
N-Chloroacetylglycine ≥99.0% (by titrimetric analysis)
N-Chloroacetylglycine
CAS Number: 6319-96-6
MDL Number: MFCD00021733
Molecular Formula: C4H6ClNO3
Molecular Weight: 151.55
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 102
Formula:
C₄H₆ClNO₃ Melting Pt: 102 °C |
MDL Number:
MFCD00021733 CAS Number: 6319-96-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...