Diethyl cyclobutane-1,1-dicarboxylate ≥97.0%

Supplier: TCI America

C0942-5ML C0942-25ML
TCC0942-5MLEA 52.14 CAD
TCC0942-5ML TCC0942-025ML
Diethyl cyclobutane-1,1-dicarboxylate ≥97.0%
Diethyl cyclobutane-1,1-dicarboxylate

CAS Number: 3779-29-1
MDL Number: MFCD00019261
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Flash Point (°C): 89
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 1,1-Cyclobutanedicarboxylic Acid Diethyl Ester

Formula: C₁₀H₁₆O₄
MW: 200.24 g/mol
Boiling Pt: 226 °C
Density: 1.05 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00019261
CAS Number: 3779-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b44d13e6556056bb5d4bd4408751e45477c4ebc&vid=4b44d13e6556056bb5d4bd4408751e45477c4ebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR