2-Cyclopentylcyclopentanone ≥99.0%

Supplier: TCI America

C0931-25ML C0931-500ML
TCC0931-025MLEA 97.96 CAD
TCC0931-025ML TCC0931-500ML
2-Cyclopentylcyclopentanone ≥99.0%
2-Cyclopentylcyclopentanone

CAS Number: 4884-24-6
MDL Number: MFCD00051419
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 233
Flash Point (°C): 104
Specific Gravity (20/20): 0.98

Formula: C₁₀H₁₆O
Boiling Pt: 233 °C
Density: 0.98 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00051419
CAS Number: 4884-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8819d989aee5205374adc7a0d7a17426775c7998&vid=8819d989aee5205374adc7a0d7a17426775c7998&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR