Vinyl cinnamate ≥99.0% stabilized
Supplier: TCI America
C0899-25ML
C0899-500ML
TCC0899-025MLEA
101.12
CAD
TCC0899-025ML
TCC0899-500ML
Vinyl cinnamate ≥99.0% stabilized
Vinyl cinnamate
(stabilized with MEHQ)
CAS Number: 3098-92-8
MDL Number: MFCD00059324
Molecular Formula: C11H10O2
Molecular Weight: 174.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Melting point (°C): -28
Flash Point (°C): 132
Specific Gravity (20/20): 1.07
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Cinnamic Acid Vinyl Ester
Formula:
C₁₁H₁₀O₂ Boiling Pt: 125 °C Melting Pt: –28 °C Density: 1.07 (20 °C) Flash Pt: 132 °C |
MDL Number:
MFCD00059324 CAS Number: 3098-92-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...