Vinyl decanoate ≥99.0% stabilized
Supplier: TCI America
Synonyms:
Decanoic Acid Vinyl Ester
C0895-25ML
C0895-500ML
TCC0895-025MLEA
88.48
CAD
TCC0895-025ML
TCC0895-500ML
Vinyl decanoate ≥99.0% stabilized
Vinyl decanoate
(stabilized with MEHQ)
CAS Number: 4704-31-8
MDL Number: MFCD00027349
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 106
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: Decanoic Acid Vinyl Ester
Formula:
C₁₂H₂₂O₂ MW: 198.31 g/mol Boiling Pt: 148 °C Density: 0.88 (20 °C) Flash Pt: 106 °C |
MDL Number:
MFCD00027349 CAS Number: 4704-31-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...