N-(2-Cyanoethyl)-N-methylaniline ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-(Methyl(phenyl)amino)propanenitrile

C0872-25G C0872-500G
TCC0872-25GEA 45.82 CAD
TCC0872-25G TCC0872-500G
N-(2-Cyanoethyl)-N-methylaniline ≥98.0% (by titrimetric analysis)
N-(2-Cyanoethyl)-N-methylaniline

CAS Number: 94-34-8
MDL Number: MFCD00019856
Molecular Formula: C10H12N2
Molecular Weight: 160.22
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 186
Flash Point (°C): 110
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: N-Methyl-N-(2-cyanoethyl)aniline, 3-(N-Methyl-N-phenylamino)propionitrile

Formula: C₁₀H₁₂N₂
MW: 160.2200 g/mol
Boiling Pt: 186 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00019856
CAS Number: 94-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b59e27cc02e48ea95e53c6aa9366c9ce5cfea9c5&vid=b59e27cc02e48ea95e53c6aa9366c9ce5cfea9c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR