Chelidamic acid monohydrate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Oxo-1,4-dihydropyridine-2,6-dicarboxylic acid monohydrate

C0821-5G C0821-25G
TCC0821-005GEA 124.82 CAD
TCC0821-005G TCC0821-025G
Chelidamic acid monohydrate ≥95.0% (by titrimetric analysis)
Chelidamic acid monohydrate

CAS Number: 138-60-3
MDL Number: MFCD00066478
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 267


Product Categories: Bioscience, Cosmetic Ingredients for Research and Experimental Use
Other Product Names / Synonyms: 4-Hydroxypyridine-2,6-dicarboxylic Acid

Formula: C₇H₅NO₅·H₂O
MW: 201.134 g/mol
Melting Pt: 267 °C
MDL Number: MFCD00066478
CAS Number: 199926-39-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85ce8b397465fecddb34860d42d99b02e7256d56&vid=f7fac9c5b77646a692e54a63a88d1d1a82f885b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR