Methyl-2-oxocyclopentanecarboxylate ≥97.0%
Supplier: TCI America
C0812-500ML
C0812-25ML
C0812-100ML
TCC0812-500MLEA
218.04
CAD
TCC0812-500ML
TCC0812-025ML
TCC0812-100ML
Methyl-2-oxocyclopentanecarboxylate ≥97.0%
Methyl 2-oxocyclopentanecarboxylate
CAS Number: 10472-24-9
MDL Number: MFCD00001411
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Flash Point (°C): 79
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: 2-Methoxycarbonylcyclopentanone, Cyclopentanone-2-carboxylic Acid Methyl Ester, Methyl Cyclopentanone-2-carboxylate, 2-Oxocyclopentanecarboxylic Acid Methyl Ester
Formula:
C₇H₁₀O₃ Boiling Pt: 111 °C Density: 1.13 (20 °C) Flash Pt: 79 °C |
MDL Number:
MFCD00001411 CAS Number: 10472-24-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...