5-Chlorosalicylamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C0787-5G C0787-25G
TCC0787-5GEA 246.32 CAD
TCC0787-5G TCC0787-025G
5-Chlorosalicylamide ≥98.0% (by HPLC, titration analysis)
5-Chlorosalicylamide

CAS Number: 7120-43-6
MDL Number: MFCD00007980
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 234


Other Product Names / Synonyms: 5-Chloro-2-hydroxybenzamide

Formula: C₇H₆ClNO₂
Melting Pt: 234 °C
MDL Number: MFCD00007980
CAS Number: 7120-43-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1241c0ed5ee92650fc769755f75c0ac4c2d8767&vid=f1241c0ed5ee92650fc769755f75c0ac4c2d8767&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR