N-Carbobenzyloxy-L-valine ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
C0761-5G
C0761-25G
TCC0761-005GEA
37.92
CAD
TCC0761-005G
TCC0761-025G
N-Carbobenzyloxy-L-valine ≥99.0% (by HPLC, titration analysis)
N-Carbobenzyloxy-L-valine
CAS Number: 1149-26-4
MDL Number: MFCD00008922
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 60
Specific rotation [a]20/D: -4.3 deg (C=2, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-valine, Z-Val-OH
Formula:
C₁₃H₁₇NO₄ Melting Pt: 60 °C |
MDL Number:
MFCD00008922 CAS Number: 1149-26-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...