Nɑ-Carbobenzoxy-L-arginine ≥97.0%

Supplier: TCI America

Synonyms: Z-Arg-OH, Nɑ-Cbz-L-arginine

C0757-1G C0757-25G
TCC0757-001GEA 56.88 CAD
TCC0757-001G TCC0757-025G
Nɑ-Carbobenzoxy-L-arginine ≥97.0%
Nalpha-Carbobenzoxy-L-arginine

CAS Number: 1234-35-1
MDL Number: MFCD00001762
Molecular Formula: C14H20N4O4
Molecular Weight: 308.34
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 173
Specific rotation [a]20/D: -10 deg (C=5.5, 0.2mol/L HCl)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-L-arginine, Z-Arg-OH

Formula: C₁₄H₂₀N₄O₄
MW: 308.34 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00001762
CAS Number: 1234-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=913210f72b3e04eb01dead18d698ad3f9e87de8a&vid=913210f72b3e04eb01dead18d698ad3f9e87de8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR