N-Carbobenzoxy-L-leucine ≥96.0%

Supplier: TCI America

Synonyms: N-Cbz-L-leucine, Z-Leu-OH

C0739-5G C0739-25G
TCC0739-005GEA 69.52 CAD
TCC0739-005G TCC0739-025G
N-Carbobenzoxy-L-leucine ≥96.0%
N-Carbobenzoxy-L-leucine

CAS Number: 2018-66-8
MDL Number: MFCD00026494
Molecular Formula: C14H19NO4
Molecular Weight: 265.31
Purity/Analysis Method: >96.0% (T)
Form: Starch Syrup
Flash Point (°C): 110
Specific rotation [a]20/D: -17 deg (C=5, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-leucine, Z-Leu-OH

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00026494
CAS Number: 2018-66-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee36680d4100640f686bd74289558285423fb299&vid=ee36680d4100640f686bd74289558285423fb299&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR