N-Carbobenzoxy-L-methionine ≥98.0%

Supplier: TCI America

Synonyms: N-Carbobenzyloxy-L-methionine, Z-Met-OH, N-Cbz-L-methionine, Cbz-Met-OH

C0737-5G C0737-25G
TCC0737-005GEA 55.3 CAD
TCC0737-005G TCC0737-025G
N-Carbobenzoxy-L-methionine ≥98.0%
N-Carbobenzoxy-L-methionine

CAS Number: 1152-62-1
MDL Number: MFCD00065124
Molecular Formula: C13H17NO4S
Molecular Weight: 283.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 68
Specific rotation [a]20/D: -26 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-methionine, Z-Met-OH

Formula: C₁₃H₁₇NO₄S
MW: 283.34 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00065124
CAS Number: 1152-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1df47e64dca8519a84b6434c8298908d58283e60&vid=1df47e64dca8519a84b6434c8298908d58283e60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR