(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Cbz-Glu-OH, Z-Glu-OH, N-Cbz-L-glutamic Acid

C0734-100G C0734-25G
TCC0734-100GEA 1030.16 CAD
TCC0734-100G TCC0734-025G
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid ≥98.0% (by titrimetric analysis)
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid

CAS Number: 1155-62-0
MDL Number: MFCD00002801
Molecular Formula: C13H15NO6
Molecular Weight: 281.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 120
Specific rotation [a]20/D: -7.5 deg (C=8, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-glutamic Acid, Z-Glu-OH

Formula: C₁₃H₁₅NO₆
MW: 281.3 g/mol
Melting Pt: 120 °C
MDL Number: MFCD00002801
CAS Number: 1155-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=373c011a7357fcad32963ea2adb661d99eaae4f0&vid=373c011a7357fcad32963ea2adb661d99eaae4f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR