N-Carbobenzoxy-L-proline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Z-Pro-OH, Z-L-Prolin, N-Cbz-L-proline, N-Carbobenzyloxy-L-proline, N-Benzyloxycarbonyl-L-proline

C0713-25G C0713-5G
TCC0713-025GEA 56.09 CAD
TCC0713-025G TCC0713-005G
N-Carbobenzoxy-L-proline ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-L-proline

CAS Number: 1148-11-4
MDL Number: MFCD00003170
Molecular Formula: C13H15NO4
Molecular Weight: 249.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 77
Specific rotation [a]20/D: -40 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-proline, Z-Pro-OH

Formula: C₁₃H₁₅NO₄
MW: 249.27 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00003170
CAS Number: 1148-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4587a15681ebaf341e12bc52612d283dad673123&vid=4587a15681ebaf341e12bc52612d283dad673123&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR