N-Carbobenzyloxy-D-alanine ≥98.0%

Supplier: TCI America

Synonyms: Z-D-Ala-OH, N-Cbz-D-alanine

C0664-25G C0664-1G C0664-5G
TCC0664-25GEA 181.7 CAD
TCC0664-25G TCC0664-001G TCC0664-005G
N-Carbobenzyloxy-D-alanine ≥98.0%
N-Carbobenzyloxy-D-alanine

CAS Number: 26607-51-2
MDL Number: MFCD00063126
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 85
Specific rotation [a]20/D: 15 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-alanine, Z-D-Ala-OH

Formula: C₁₁H₁₃NO₄
MW: 223.2 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00063126
CAS Number: 26607-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd635fecfdfaddd66611967376471d471a25f354&vid=bd635fecfdfaddd66611967376471d471a25f354&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR