N-Carbobenzoxy-D-phenylalanine ≥98.0%
Supplier: TCI America
Synonyms:
N-Cbz-D-phenylalanine, Z-D-Phe-OH, ((Benzyloxy)carbonyl)-D-phenylalanine, Cbz-D-Phe-OH
C0662-5G
C0662-1G
TCC0662-5GEA
210.14
CAD
TCC0662-5G
TCC0662-001G
N-Carbobenzoxy-D-phenylalanine ≥98.0%
N-Carbobenzoxy-D-phenylalanine
CAS Number: 2448-45-5
MDL Number: MFCD00063151
Molecular Formula: C17H17NO4
Molecular Weight: 299.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 88
Specific rotation [a]20/D: -5.3 deg (C=4, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-phenylalanine, Z-D-Phe-OH
Formula:
C₁₇H₁₇NO₄ Melting Pt: 88 °C |
MDL Number:
MFCD00063151 CAS Number: 2448-45-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...