(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid ≥98.0%
Supplier: TCI America
Synonyms:
Cbz-Phe-OH, N-Cbz-L-phenylalanine, N-Carbobenzyloxy-L-phenylalanine, Z-Phe-OH
C0660-5G
C0660-25G
TCC0660-005GEA
36.34
CAD
TCC0660-005G
TCC0660-025G
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid ≥98.0%
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
CAS Number: 1161-13-3
MDL Number: MFCD00020418
Molecular Formula: C17H17NO4
Molecular Weight: 299.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 86
Specific rotation [a]20/D: 5.3 deg (C=4, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-phenylalanine, Z-Phe-OH
Formula:
C17H17NO4 Melting Pt: 86 °C |
MDL Number:
MFCD00020418 CAS Number: 1161-13-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...