trans-m-Coumaric acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

C0655-25G C0655-5G
TCC0655-025GEA 83.74 CAD
TCC0655-025G TCC0655-5G
trans-m-Coumaric acid ≥99.0% (by GC, titration analysis)
trans-m-Coumaric acid

CAS Number: 14755-02-3
MDL Number: MFCD00004390
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 196


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: trans-m-Cumaric Acid, trans-3-Hydroxycinnamic Acid

Formula: C₉H₈O₃
Melting Pt: 196 °C
MDL Number: MFCD00004390
CAS Number: 14755-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63679b6f7fdb861a469d702e2115655986a7f939&vid=26e24dc752c27cbdacc993d8308fd91677346d36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR