N-Cbz-L-serine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine
C0635-25G
C0635-5G
TCC0635-25GEA
137.46
CAD
TCC0635-25G
TCC0635-5G
N-Cbz-L-serine ≥98.0% (by HPLC, titration analysis)
N-Cbz-L-serine
CAS Number: 1145-80-8
MDL Number: MFCD00002662
Molecular Formula: C11H13NO5
Molecular Weight: 239.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 118
Specific rotation [a]20/D: 6.1 deg (C=6, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-serine, Z-Ser-OH
Formula:
C₁₁H₁₃NO₅ MW: 239.2 g/mol Melting Pt: 118 °C |
MDL Number:
MFCD00002662 CAS Number: 1145-80-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...