N-Carbobenzyloxy-L-alanine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine

C0633-10G C0633-25G
TCC0633-010GEA 42.66 CAD
TCC0633-010G TCC0633-025G
N-Carbobenzyloxy-L-alanine ≥99.0% (by HPLC, titration analysis)
N-Carbobenzyloxy-L-alanine

CAS Number: 1142-20-7
MDL Number: MFCD00002640
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 86
Specific rotation [a]20/D: -14.5 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-alanine, Z-Ala-OH

Formula: C11H13NO4
MW: 223.23 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00002640
CAS Number: 1142-20-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a49dd091c30bf86514a4523f57329b6a91f2bfdc&vid=a49dd091c30bf86514a4523f57329b6a91f2bfdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR