N-Carbobenzoxy-L-aspartic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Cbz-L-aspartic Acid, N-Carbobenzyloxy-L-aspartic acid, Z-Asp-OH

C0629-25G C0629-5G
TCC0629-25GEA 107.66 CAD
TCC0629-25G TCC0629-5G
N-Carbobenzoxy-L-aspartic acid ≥99.0% (by titrimetric analysis)
N-Carbobenzoxy-L-aspartic acid

CAS Number: 1152-61-0
MDL Number: MFCD00002719
Molecular Formula: C12H13NO6
Molecular Weight: 267.24
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 116
Specific rotation [a]20/D: 9.5 deg (C=7, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-aspartic Acid, Z-Asp-OH

Formula: C₁₂H₁₃NO₆
MW: 267.24 g/mol
Melting Pt: 116 °C
MDL Number: MFCD00002719
CAS Number: 1152-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a93c20119754343ec99a992938f185a89923f9c5&vid=a93c20119754343ec99a992938f185a89923f9c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR