2'-Chloroacetoacetanilide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C0624-500G C0624-25G
TCC0624-500GEA 213.3 CAD
TCC0624-500G TCC0624-025G
2'-Chloroacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
2'-Chloroacetoacetanilide

CAS Number: 93-70-9
MDL Number: MFCD00018224
Molecular Formula: C10H10ClNO2
Molecular Weight: 211.65
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 107


Other Product Names / Synonyms: N-Acetoacetyl-2-chloroaniline

Formula: C₁₀H₁₀ClNO₂
Melting Pt: 107 °C
MDL Number: MFCD00018224
CAS Number: 93-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73d4a42c90e6ad8a249e33dd3f0bdeb378c5b46a&vid=73d4a42c90e6ad8a249e33dd3f0bdeb378c5b46a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR