N-Cbz-L-asparagine 99,0

Supplier: TCI America

Synonyms: Z-Asn-OH, Nalpha-Cbz-L-asparagine

C0573-1G C0573-10G
TCC0573-001GEA 52.14 CAD
TCC0573-001G TCC0573-010G
N-Cbz-L-asparagine 99,0
N-Cbz-L-asparagine

CAS Number: 2304-96-3
MDL Number: MFCD00008035
Molecular Formula: C12H14N2O5
Molecular Weight: 266.25
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Specific rotation [a]20/D: 6.3 deg (C=1.6, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-L-asparagine, Z-Asn-OH

Formula: C₁₂H₁₄N₂O₅
MW: 266.3 g/mol
MDL Number: MFCD00008035
CAS Number: 2304-96-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f379ff13505fda109e779cf2accbcbf8aa5da30&vid=8f379ff13505fda109e779cf2accbcbf8aa5da30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR