Carmine 45.0% (by HPLC)
Supplier: TCI America
C0543-5G
C0543-25G
TCC0543-005GEA
61.62
CAD
TCC0543-005G
TCC0543-025G
Carmine 45.0% (by HPLC)
Carmine
CAS Number: 1390-65-4
MDL Number: MFCD00167028
Purity/Analysis Method: >40.0% (HPLC)
Form: Crystal
Other Product Names / Synonyms: Carmine Red
Formula:
C22H20O13 |
MDL Number:
MFCD00167028 CAS Number: 1390-65-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...