Cyclohexylamine hydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

C0495-500G C0495-25G
TCC0495-500GEA 184.86 CAD
TCC0495-500G TCC0495-025G
Cyclohexylamine hydrochloride ≥98.0% (by total nitrogen basis)
Cyclohexylamine hydrochloride

CAS Number: 4998-76-9
MDL Number: MFCD00050540
Molecular Formula: C6H13N
Molecular Weight: 135.64
Purity/Analysis Method: >98.0% (N)
Form: Crystal

Formula: C₆H₁₃N
MDL Number: MFCD00050540
CAS Number: 4998-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c1f5e919be24259d7457ac0782bf9abd0ce3987&vid=1c1f5e919be24259d7457ac0782bf9abd0ce3987&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR