cis-4-Cyclohexene-1,2-dicarboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
(1R,2S)-rel-Cyclohex-4-ene-1,2-dicarboxylic acid
C0492-25G
C0492-500G
TCC0492-025GEA
50.56
CAD
TCC0492-025G
TCC0492-500G
cis-4-Cyclohexene-1,2-dicarboxylic acid ≥98.0%
cis-4-Cyclohexene-1,2-dicarboxylic acid
CAS Number: 2305-26-2
MDL Number: MFCD00070482
Molecular Formula: C8H10O4
Molecular Weight: 170.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 168
Flash Point (°C): 200
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: cis-DELTA4-Tetrahydrophthalic Acid
Formula:
C₈H₁₀O₄ Melting Pt: 168 °C Flash Pt: 200 °C |
MDL Number:
MFCD00070482 CAS Number: 2305-26-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...