cis-1,2-Cyclohexanedicarboxylic anhydride ≥99.0%

Supplier: TCI America

C0478-500G C0478-25G C0478-100G
TCC0478-500GEA 115.34 CAD
TCC0478-500G TCC0478-025G TCC0478-100G
cis-1,2-Cyclohexanedicarboxylic anhydride ≥99.0%
cis-1,2-Cyclohexanedicarboxylic anhydride

CAS Number: 13149-00-3
MDL Number: MFCD00005926
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 160
Flash Point (°C): 153
Freezing point (°C): 36

Formula: C₈H₁₀O₃
Boiling Pt: 160 °C
Flash Pt: 153 °C
MDL Number: MFCD00005926
CAS Number: 13149-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e2ce3c8375e94c207769936a2153e20383e5a59&vid=2e2ce3c8375e94c207769936a2153e20383e5a59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR