trans-4-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C0393-500G
C0393-25G
C0393-100G
TCC0393-500GEA
813.23
CAD
TCC0393-500G
TCC0393-025G
TCC0393-100G
trans-4-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-4-Hydroxycinnamic acid
CAS Number: 501-98-4
MDL Number: MFCD00004399
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 213
Product Categories: Bioscience, Antioxidants, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: trans-p-Cumaric Acid, trans-4-Hydroxycinnamic Acid
Formula:
C₉H₈O₃ Melting Pt: 213 °C |
MDL Number:
MFCD00004399 CAS Number: 501-98-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...