2-Chloro-p-phenylenediamine sulfate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2-Chlorobenzene-1,4-diamine sulphate, 2-Chloro-1,4-phenylenediammonium sulfate, 2-Chloro-1,4-phenylenediammonium sulphate, 2-Chlorobenzene-1,4-diamine sulfate

C0256-500G C0256-25G
TCC0256-500GEA 1112.32 CAD
TCC0256-500G TCC0256-25G
2-Chloro-p-phenylenediamine sulfate ≥98.0% (by HPLC, total nitrogen)
2-Chloro-p-phenylenediamine sulphate

CAS Number: 61702-44-1
MDL Number: MFCD00013004
Molecular Formula: C6H7ClN2
Molecular Weight: 240.66
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 253


Other Product Names / Synonyms: 1,4-Diamino-2-chlorobenzene Sulfate

Formula: C₆H₉ClN₂O₄S
MW: 240.67 g/mol
Melting Pt: 253 °C
MDL Number: MFCD00013004
CAS Number: 61702-44-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a027162a78bbb9f5b98fc384dc3c9a6fcb2f9929&vid=a027162a78bbb9f5b98fc384dc3c9a6fcb2f9929&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR