4-Chloro-DL-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: p-Chloro-DL-Phenylalanine, H-DL-Phe(4-Cl)-OH

C0253-25G C0253-1G
TCC0253-025GEA 459.78 CAD
TCC0253-025G TCC0253-001G
4-Chloro-DL-phenylalanine ≥98.0%
4-Chloro-DL-phenylalanine

CAS Number: 7424-00-2
MDL Number: MFCD00002601
Molecular Formula: C9H10ClNO2
Molecular Weight: 199.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Amino-3-(4-chlorophenyl)propionic Acid, H-DL-Phe(4-Cl)-OH

Formula: C₉H₁₀ClNO₂
MW: 199.6 g/mol
MDL Number: MFCD00002601
CAS Number: 7424-00-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bc7f2ece26d920332cdc0924ac3391cf0f91e45&vid=6bc7f2ece26d920332cdc0924ac3391cf0f91e45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR