Allyl chloroacetate ≥98.0%

Supplier: TCI America

Synonyms: Chloroacetic acid allyl ester, Allyl 2-chloroacetate

C0090-500G C0090-25G
TCC0090-500GEA 1025.42 CAD
TCC0090-500G TCC0090-25G
Allyl chloroacetate ≥98.0%
Allyl chloroacetate

CAS Number: 2916-14-5
MDL Number: MFCD00058936
Molecular Formula: C5H7ClO2
Molecular Weight: 134.56
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 61
Specific Gravity (20/20): 1.16


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Chloroacetic Acid Allyl Ester

Formula: C₅H₇ClO₂
MW: 134.56 g/mol
Boiling Pt: 164 °C
Density: 1.16 (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00058936
CAS Number: 2916-14-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=119a12962eef681ea57034713290893773a8fdb9&vid=119a12962eef681ea57034713290893773a8fdb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR