Carboxymethyl cellulose sodium salt n=∼ 500

Supplier: TCI America

Synonyms: Carboxymethyl cellulose, sodium saltCellulose, carboxymethyl ether, sodium saltCMCCellulose gumSodium carboxymethyl celluloseSodium carboxymethyl cellulose (Viscosity 800mpa.s)

C0045-500G C0045-25G
TCC0045-500GEA 83.44 CAD
TCC0045-500G TCC0045-025G
Carboxymethyl cellulose sodium salt n=∼ 500
Carboxymethyl cellulose sodium salt

CAS Number: 9004-32-4
MDL Number: MFCD00081472
Form: Crystal
Color: White


Other Product Names / Synonyms: Cellulose Glycolic Acid Sodium Salt, Sodium Carboxymethyl Cellulose, Sodium Cellulose Glycolate, Sodium Tylose, Tylose Sodium

Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number: MFCD00081472
CAS Number: 9004-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR