Guaiacol carbonate ≥98.0% (by HPLC)

Supplier: TCI America

C0039-500G C0039-25G
TCC0039-500GEA 929.04 CAD
TCC0039-500G TCC0039-025G
Guaiacol carbonate ≥98.0% (by HPLC)
Guaiacol carbonate

CAS Number: 553-17-3
MDL Number: MFCD00025723
Molecular Formula: C15H14O5
Molecular Weight: 274.27
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 88


Other Product Names / Synonyms: Bis(2-methoxyphenyl) Carbonate, Carbonic Acid Bis(2-methoxyphenyl) Ester

Formula: C₁₅H₁₄O₅
Melting Pt: 88 °C
MDL Number: MFCD00025723
CAS Number: 553-17-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR