Nε-carbobenzoxy-L-lysine ≥98.0%

Supplier: TCI America

Synonyms: Nε-Cbz-L-lysine, (S)-2-Amino-6-(((benzyloxy)carbonyl)amino)hexanoic acid, H-Lys(Z)-OH

C0034-5G C0034-25G
TCC0034-005GEA 90.06 CAD
TCC0034-005G TCC0034-025G
Nε-carbobenzoxy-L-lysine ≥98.0%
Nepsilon-carbobenzoxy-L-lysine

CAS Number: 1155-64-2
MDL Number: MFCD00002638
Molecular Formula: C14H20N2O4
Molecular Weight: 280.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 16 deg (C=1.6, 2mol/L HCl)


Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: Nepsilon-Cbz-L-lysine, H-Lys(Z)-OH

Formula: C₁₄H₂₀N₂O₄
MW: 280.32 g/mol
MDL Number: MFCD00002638
CAS Number: 1155-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2488fb835725870be98f3c68f1f713aa7752d547&vid=2488fb835725870be98f3c68f1f713aa7752d547&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR