(1R,3S)-(+)-Camphoric acid ≥98.0%
Supplier: TCI America
Synonyms:
D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B
C0012-25G
C0012-100G
TCC0012-025GEA
134.3
CAD
TCC0012-025G
TCC0012-100G
(1R,3S)-(+)-Camphoric acid ≥98.0%
(1R,3S)-(+)-Camphoric acid
CAS Number: 124-83-4
MDL Number: MFCD00001375
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 187
Specific rotation [a]20/D: 46 deg (C=5, EtOH)
Formula:
C₁₀H₁₆O₄ MW: 200.23 g/mol Melting Pt: 187 °C |
MDL Number:
MFCD00001375 CAS Number: 124-83-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...