Calmagite
Supplier: TCI America
C0008-1G
C0008-5G
TCC0008-001GEA
116.92
CAD
TCC0008-001G
TCC0008-005G
Calmagite
Calmagite
[Metal indicator for Ca, Mg etc.]
CAS Number: 3147-14-6
MDL Number: MFCD00011656
Molecular Formula: C17H14N2O5S
Molecular Weight: 358.37
Form: Crystal
Color: Deep Yellow Red
Melting point (°C): 330
Lambda max.: 526 nm (H2O)
Other Product Names / Synonyms: 1-(1-Hydroxy-4-methyl-2-phenylazo)-2-naphthol-4-sulfonic Acid
Formula:
C₁₇H₁₄N₂O₅S Melting Pt: 330 °C |
MDL Number:
MFCD00011656 CAS Number: 3147-14-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...