N-Boc-azetidine-3-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: Boc-azetidine-3-carboxylic acid, N-Boc-Azetidine-3-carboxylic acid, 1-N-Boc-3-Azetidinecarboxylic acid, 1-Boc-azetidine-3-carboxylic Acid

B3540-1G B3540-5G
TCB3540-1GEA 143.1 CAD
TCB3540-1G TCB3540-5G
N-Boc-azetidine-3-carboxylic acid ≥98.0%
N-Boc-azetidine-3-carboxylic acid

CAS Number: 142253-55-2
MDL Number: MFCD01860897
Molecular Formula: C9H15NO4
Molecular Weight: 201.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 105


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-Boc-azetidine-3-carboxylic Acid

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
Melting Pt: 105 °C
MDL Number: MFCD01860897
CAS Number: 142253-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2a2b7089400ea24de82b2979f3d09f968d39b1a&vid=a2a2b7089400ea24de82b2979f3d09f968d39b1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR