2-Bromoquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3524-1G B3524-5G
TCB3524-1GEA 67.78 CAD
TCB3524-1G TCB3524-5G
2-Bromoquinoline ≥98.0% (by GC, titration analysis)
2-Bromoquinoline

CAS Number: 2005-43-8
MDL Number: MFCD00234480
Molecular Formula: C9H6BrN
Molecular Weight: 208.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 49
Flash Point (°C): 110
Lambda max.: 319 nm (EtOH)

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 115 °C
Melting Pt: 49 °C
Flash Pt: 110 °C
MDL Number: MFCD00234480
CAS Number: 2005-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53c0158ac0640eee6872ffd84d2dae4301e4911e&vid=76e3485a6e6c81ee1f870653114b36f794c9ec9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR