tert-Butyl 2-bromoisobutyrate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: tert-Butyl α-bromoisobutyrate, 2-Bromoisobutyric acid tert-butyl ester, tert-Butyl 2-methyl-2-propionate, tert-Butyl 2-bromo-2-methylpropanoate

B3500-25G B3500-500G
TCB3500-25GEA 73.33 CAD
TCB3500-25G TCB3500-500G
tert-Butyl 2-bromoisobutyrate ≥98.0% (by GC)
tert-Butyl 2-bromoisobutyrate

CAS Number: 23877-12-5 MDL Number: MFCD00051908 Molecular Formula: C8H15BrO2 Molecular Weight: 223.11 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 37 Specific Gravity (20/20): 1.20


Product Categories: Materials Chemistry, Precise Polymerization Reagents Other Product Names / Synonyms: 2-Bromoisobutyric Acid tert-Butyl Ester

Formula: C₆H₁₁BrO₂
MW: 223.12 g/mol
Boiling Pt: 37 °C (1mm Hg) (lit.)
Density: 1.20 (20 °C)
MDL Number: MFCD00051908
CAS Number: 23877-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8eec1d9727792e44f06d765cf53de09cccb904a&vid=b8eec1d9727792e44f06d765cf53de09cccb904a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR