2-Bromo-1,3-dimethoxybenzene ≥98.0%

Supplier: TCI America

B3488-5G B3488-1G B3488-25G
TCB3488-5GEA 191.18 CAD
TCB3488-5G TCB3488-1G TCB3488-25G
2-Bromo-1,3-dimethoxybenzene ≥98.0%
2-Bromo-1,3-dimethoxybenzene

CAS Number: 16932-45-9
MDL Number: MFCD07780177
Molecular Formula: C8H9BrO2
Molecular Weight: 217.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 273
Melting point (°C): 95

Formula: C₈H₉BrO₂
Boiling Pt: 273 °C
Melting Pt: 95 °C
MDL Number: MFCD07780177
CAS Number: 16932-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1b3e51055b795b93b97f9cd58a5046ea3ba8613&vid=e1b3e51055b795b93b97f9cd58a5046ea3ba8613&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR