3-Bromo-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3482-5G B3482-25G
TCB3482-5GEA 243.32 CAD
TCB3482-5G TCB3482-25G
3-Bromo-o-toluidine ≥98.0% (by GC, titration analysis)
3-Bromo-o-toluidine

CAS Number: 55289-36-6
MDL Number: MFCD00051579
Molecular Formula: C7H8BrN
Molecular Weight: 186.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 255
Flash Point (°C): 110
Specific Gravity (20/20): 1.52


Other Product Names / Synonyms: 2-Amino-6-bromotoluene, 3-Bromo-o-toluidine

Formula: C₇H₈BrN
Boiling Pt: 255 °C
Density: 1.52 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00051579
CAS Number: 55289-36-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c53ffb8292c3fb3af82302c05c1b2b1d1108c29b&vid=c53ffb8292c3fb3af82302c05c1b2b1d1108c29b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR