2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0%

Supplier: TCI America

B3448-5G B3448-1G
TCB3448-5GEA 193.39 CAD
TCB3448-5G TCB3448-1G
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0%
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

CAS Number: 87100-28-5
MDL Number: MFCD05663841
Molecular Formula: C13H19BO2
Molecular Weight: 218.10
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 65
Flash Point (°C): 110
Specific Gravity (20/20): 0.99


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Benzylboronic Acid Pinacol Ester

Formula: C₁₃H₁₉BO₂
Boiling Pt: 65 °C
Density: 0.99 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD05663841
CAS Number: 87100-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21707bdc28824fd9d3975e478bda173927910fd7&vid=21707bdc28824fd9d3975e478bda173927910fd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR