4-Bromo-3-nitroanisole ≥96.0%
Supplier: TCI America
B3420-25G
B3420-5G
TCB3420-25GEA
209.98
CAD
TCB3420-25G
TCB3420-5G
4-Bromo-3-nitroanisole ≥96.0%
4-Bromo-3-nitroanisole
CAS Number: 5344-78-5
MDL Number: MFCD00051511
Molecular Formula: C7H6BrNO3
Molecular Weight: 232.03
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 32
Flash Point (°C): 113
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-Bromo-4-methoxy-2-nitrobenzene
Formula:
C₇H₆BrNO₃ Boiling Pt: 154 °C Melting Pt: 32 °C Flash Pt: 113 °C |
MDL Number:
MFCD00051511 CAS Number: 5344-78-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...